Learn More
Ethyl 3-oxo-3-(pyridin-2-yl)propanoate, 96%, BLD Pharm™
Specifications
Specifications
| CAS | 26510-52-1 |
| Molecular Formula | C10H11NO3 |
| MDL Number | MFCD00094022 |
| Quantity | 250 mg |
| SMILES | O=C(OCC)CC(C1=NC=CC=C1)=O |
| Molecular Weight (g/mol) | 193.20 |
| Percent Purity | 96% |
| Chemical Name or Material | Ethyl 3-oxo-3-(pyridin-2-yl)propanoate |