Learn More
Ethyl 3-Hydroxy-3-methyl-d3-butyrate-4,4,4-d3, CDN
Specifications
Specifications
| Chemical Name or Material | 3-Hydroxy-3-methylbutanoic Acid Ethyl Ester-4,4,4,4',4',4'-D6 |
| CAS | 30385-86-5 |
| Molecular Formula | C7 D6 H8 O3 |
| Quantity | 0.5 g |
| Synonym | Butanoic-4,4,4-d3 acid, 3-hydroxy-3-(methyl-d3)-, ethyl ester (9 CI), Butyric-4,4,4-d3 acid, 3-hydroxy-3-methyl-d3-, ethyl ester (8 CI), Ethyl 3-Hydroxy-3-methyl-d3-butyrate-4,4,4-d3, Ethyl 3-hydroxy-3-(methyl-d3)butanoate-4,4,4-d3, Ethyl 3-hydroxy-3-(methyl-d3)butyrate-4,4,4-d3, 3-Hydroxy-3-methylbutanoic Acid Ethyl Ester-4,4,4,4',4',4'-D6, 3-Hydroxy-3-methylbutanoic Acid Ethyl Ester-4,4,4,4',4',4'-D6 |
| InChI Formula | InChI=1 S/C7H14O3/c1-4-10-6(8)5-7(2,3)9/h9H,4-5H2,1-3H3/i2D3,3D3 |
| SMILES | [2 H]C([2 H])([2 H])C(O)(CC(=O)OCC)C([2 H])([2 H])[2 H] |
| IUPAC Name | ethyl 4,4,4-trideuterio-3-hydroxy-3-(trideuteriomethyl)butanoate |
| Molecular Weight (g/mol) | 152.2212 |
| Formula Weight | 152.132 g/mol |
| Show More |