Learn More
Ethyl 3-Aminobutanoate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl 3-Aminobutanoate |
| CAS | 5303-65-1 |
| Molecular Formula | C6H13NO2 |
| Quantity | 50 mg |
| Synonym | 3-Aminobutanoic Acid Ethyl Ester, 3-Aminobutyric Acid Ethyl Ester, 3-Methyl-beta-alanine Ethyl Ester, Ethyl 3-aminobutyrate, Ethyl beta-aminobutyrate, NSC 77084, [2-(Ethoxycarbonyl)-1-methylethyl]amine, beta-Aminobutyric Acid Ethyl Ester |
| InChI Formula | InChI=1S/C6H13NO2/c1-3-9-6(8)4-5(2)7/h5H,3-4,7H2,1-2H3 |
| SMILES | CCOC(=O)CC(C)N |
| IUPAC Name | ethyl 3-aminobutanoate |
| Molecular Weight (g/mol) | 131.17 |
| Formula Weight | 131.0946 |