Learn More
Ethyl 3-(4-Chlorophenyl)-3-oxopropanoate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl 3-(4-Chlorophenyl)-3-oxopropanoate |
| CAS | 2881-63-2 |
| Molecular Formula | C11H11O3Cl |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C11H11ClO3/c1-2-15-11(14)7-10(13)8-3-5-9(12)6-4-8/h3-6H,2,7H2,1H3 |
| SMILES | CCOC(=O)CC(=O)c1ccc(Cl)cc1 |
| IUPAC Name | ethyl 3-(4-chlorophenyl)-3-oxopropanoate |
| Molecular Weight (g/mol) | 226.65 |
| Formula Weight | 226.0397 |