Learn More
Ethyl 2-chloro-5-pyridylglyoxylate, 97%, Thermo Scientific Chemicals
Product Code. 11439280
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439280 1 g 1g
11449280 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11439280 Supplier Thermo Scientific Alfa Aesar Supplier No. H50157.03
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
CAS: 902837-55-2 | Molecular Formula: C9H8ClNO3 | Molecular Weight: 213.617 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 902837-55-2
Molecular Formula C9H8ClNO3
Molecular Weight (g/mol) 213.617
MDL Number MFCD07781188
InChI Key HRFPVENOVYSKHE-UHFFFAOYSA-N
Synonym ethyl 6-chloro-3-pyridylglyoxylate, ethyl 2-6-chloropyridin-3-yl-2-oxoacetate, ethyl 2-chloro-5-pyridylglyoxylate, ethyl 6-chloropyridin-3-yl oxo acetate, ethyl6-chloro-3-pyridylglyoxylate, ethyl 6-chloro-3-pyridinyl oxoacetate, 6-chloro-3-pyridylglyoxylic acid ethyl ester, ethyl 2-6-chloro 3-pyridyl-2-oxoacetate
PubChem CID 42553168
IUPAC Name ethyl 2-(6-chloropyridin-3-yl)-2-oxoacetate
SMILES CCOC(=O)C(=O)C1=CN=C(C=C1)Cl
Specifications
Quantity 1 g
Formula Weight 213.62
Percent Purity 97%
Chemical Name or Material Ethyl 2-chloro-5-pyridylglyoxylate