Learn More
Ethyl 2-amino-4-phenylthiazole-5-carboxylate, 98%, Thermo Scientific™
Product Code. 11383287
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11383287
Brand: Thermo Scientific Alfa Aesar L13864.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 64399-23-1
Molecular Formula C12H12N2O2S
Molecular Weight (g/mol) 248.3
MDL Number MFCD00022452
InChI Key OZMXFXOHCUEEPD-UHFFFAOYSA-N
PubChem CID 73239
IUPAC Name ethyl 2-amino-4-phenyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=C(S1)N)C2=CC=CC=C2
Specifications
Melting Point 170°C to 173°C
Quantity 5 g
Beilstein 222259
Synonym ethyl 2-amino-4-phenylthiazole-5-carboxylate, ethyl 2-amino-4-phenyl-5-thiazolecarboxylate, 5-thiazolecarboxylic acid, 2-amino-4-phenyl-, ethyl ester, 2-amino-4-phenylthiazole-5-carboxylic acid ethyl ester, acmc-209nlv, 2-amino-4-phenyl-5-carboethoxythiazole, ethyl 2-amino-4-phenylthiazol-5-carboxylate, ethyl2-amino-4-phenylthiazole-5-carboxylate, ethyl 2-amino-4-phenyl-thiazole-5-carboxylate, ethyl 2-amino-4-phenylthiazol-5-yl-carboxylate
Formula Weight 248.31
Percent Purity 98%
Chemical Name or Material Ethyl 2-amino-4-phenylthiazole-5-carboxylate