Learn More
Ethyl 2-(4-(trifluoromethyl)phenyl)acetate, 98%, BLD Pharm™
Product Code. 31430590
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
100 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31430590 25 g
31420465 10 g
31420877 5 g
31425384 1 g
31427616 250 mg
31423898 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31430590 Supplier BLD Pharm Supplier No. BD1299725g
 Restricted Item
Estimated Shipment: 25-11-2026
Specifications
CAS 721-63-1
Molecular Formula C11H11F3O2
MDL Number MFCD07779351
Quantity 25 g
SMILES O=C(OCC)CC1=CC=C(C(F)(F)F)C=C1
Molecular Weight (g/mol) 232.20
Percent Purity 98%
Chemical Name or Material Ethyl 2-(4-(trifluoromethyl)phenyl)acetate