Learn More
Ethyl 2,4-Dioxovalerate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl 2,4-Dioxovalerate |
| CAS | 615-79-2 |
| Molecular Formula | C7H10O4 |
| Quantity | 10 g |
| Synonym | 2,4-Dioxovaleric Acid Ethyl Ester, 2,4-Dioxopentanoic Acid Ethyl Ester, Ethoxalylacetone, Ethyl (Acetylmethyl)glyoxylate, Ethyl 2,4-Dioxopentanoate, Ethyl Acetopyruvate, Ethyl Acetylpyruvate, NSC 1243, NSC 97434 |
| InChI Formula | InChI=1S/C7H10O4/c1-3-11-7(10)6(9)4-5(2)8/h3-4H2,1-2H3 |
| SMILES | CCOC(=O)C(=O)CC(=O)C |
| IUPAC Name | ethyl 2,4-dioxopentanoate |
| Molecular Weight (g/mol) | 158.15 |
| Formula Weight | 158.0579 |