Learn More
Ethyl-2,2-diethoxyacetate, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Ethyl-2,2-diethoxyacetate |
| CAS | 6065-82-3 |
| Molecular Formula | C8 H16 O4 |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C8H16O4/c1-4-10-7(9)8(11-5-2)12-6-3/h8H,4-6H2,1-3H3 |
| SMILES | CCOC(OCC)C(=O)OCC |
| IUPAC Name | ethyl 2,2-diethoxyacetate |
| Molecular Weight (g/mol) | 176.21 |
| Formula Weight | 176.1049 |