Learn More
Ethyl-1,2,2,2-d4 Alcohol, CDN
Product Code. 30321328
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321328
Brand: CDN CDND54750.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ethanol-1,2,2,2-D4
CAS 22544-43-0
Molecular Formula C2 D4 H2 O
Quantity 0.5 g
Synonym Ethan-1,2,2,2-d4-ol (9 CI), Ethyl-1,2,2,2-d4 alcohol (8 CI), Ethanol-1,2,2,2-D4, Ethanol-d4
InChI Formula InChI=1 S/C2H6O/c1-2-3/h3H,2H2,1H3/i1D3,2 D
SMILES [2 H]C(O)C([2 H])([2 H])[2 H]
IUPAC Name 1,2,2,2-tetradeuterioethanol
Molecular Weight (g/mol) 50.0931
Formula Weight 50.067 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity