missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethoxyquin-d5 HCl (ethyl-d5), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Ethoxyquin D5 (Ethyl D5) Hydrochloride |
| Molecular Formula | C14 D5 H14 N O . Cl H |
| Quantity | 0.01 g |
| Synonym | Ethoxyquin D5 (Ethyl D5) Hydrochloride, 6-(Ethoxy-d5)-2,2,4-trimethyl-1,2-dihydroquinoline hydrochloride |
| InChI Formula | InChI=1 S/C14H19NO.ClH/c1-5-16-11-6-7-13-12(8-11)10(2)9-14(3,4)15-13;/h6-9,15 H,5H2,1-4H3;1 H/i1D3,5D2; |
| SMILES | Cl.[2 H]C([2 H])([2 H])C([2 H])([2 H])Oc1ccc2NC(C)(C)C=C(C)c2c1 |
| IUPAC Name | 2,2,4-trimethyl-6-(1,1,2,2,2-pentadeuterioethoxy)-1 H-quinoline;hydrochloride |
| Molecular Weight (g/mol) | 258.7985 |
| Formula Weight | 258.1547 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction