Learn More
Draeger Safety Ethanol Diffusion Tube
Product Code. 15613176
Change view
Click to view available options
Components Measured:
Ethanol 1000/a-D
Unit Size:
Pack of 10
Product Code. Components Measured unitSize
15613176 Ethanol 1000/a-D Pack of 10
1 options
Product Code. 15613176
Brand: Draeger Safety 8101151
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Draeger Safety Ethanol Diffusion Tube is a direct-reading diffusion tube used specifically for personal exposure monitoring.
  • Direct-reading diffusion tubes are used specifically for personal exposure monitoring
  • These tubes do not require the use of a gas detector pump because the contaminant molecules pass into the tube by diffusion processes and in this manner are brought into contact with the reagent systems
  • Exposure levels are easily read by the markings on the side of the tube
Specifications
Components Measured Ethanol 1000/a-D
Type Diffusion Tube
Measurement Range 1 hr: 1000 to 25,000 ppm, 4 hr: 125 to 3100 ppm