missing translation for 'onlineSavingsMsg'
Learn More

Ethanesulfonic acid, 97%, Thermo Scientific™

Product Code. p-7043713
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11341567

Brand: Thermo Scientific Alfa Aesar L02270.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethanesulfonic acid is used as pharmaceutical intermediates, catalyst of polymerization and alkylation. It participates in electrolytic reduction of perrhenate solutions.

Solubility
Completely soluble in water.

Notes
Store at room temperature. Incompatible with Strong oxidizing agents, Strong bases. Store away from strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 594-45-6
Molecular Formula C2H5O3S
Molecular Weight (g/mol) 109.12
MDL Number MFCD00007529
InChI Key CCIVGXIOQKPBKL-UHFFFAOYSA-M
Synonym ethylsulfonic acid, ethanesulphonic acid, ethane sulfonate, ethane sulfonic acid, ethane sulphonic acid, ethansulfonic acid, ethylsulphonic acid, 1-ethanesulfonic acid, 2-ethanesulfonic acid, ethanesulfonicacid
PubChem CID 11668
ChEBI CHEBI:42465
IUPAC Name ethanesulfonic acid
SMILES CCS([O-])(=O)=O

Specifications

Melting Point -17°C
Boiling Point 122°C to 123°C (0.01 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
UN Number UN2586
Beilstein 1743071
Formula Weight 110.13
Percent Purity 97%
Chemical Name or Material Ethanesulfonic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.