missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-4-aminophenethylamine, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | N-Boc-4-aminophenethylamine |
| CAS | 94838-59-2 |
| Molecular Formula | C13H20N2O2 |
| Quantity | 2.5 g |
| Synonym | Carbamic acid, [2-(4-aminophenyl)ethyl]-, 1,1-dimethylethyl ester (9CI), N-tert-Butoxycarbonyl-2-(4-aminophenyl)ethylamine, N-tert-Butoxycarbonyl-4-aminophenethylamine, [2-(4-Aminophenyl)ethyl]carbamic acid tert-butyl ester, tert-Butyl (2-(4-Aminophenyl)ethyl)carbamate, tert-Butyl 4-aminophenethylcarbamate |
| InChI Formula | InChI=1S/C13H20N2O2/c1-13(2,3)17-12(16)15-9-8-10-4-6-11(14)7-5-10/h4-7H,8-9,14H2,1-3H3,(H,15,16) |
| SMILES | CC(C)(C)OC(=O)NCCc1ccc(N)cc1 |
| IUPAC Name | tert-butyl N-[2-(4-aminophenyl)ethyl]carbamate |
| Molecular Weight (g/mol) | 236.31 |
| Formula Weight | 236.1525 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction