Learn More
Estriol, TRC
Product Code. 18061730
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18061730 1 g 1g
18011040 500 mg 500mg
18062120 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18061730 Supplier Toronto Research Chemicals Supplier No. TRCE8889601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 50-27-1
Molecular Formula C18 H24 O3
Molecular Weight (g/mol) 288.38
Synonym Estriol, (16α,17β)-Estra-1,3,5(10)-triene-3,16,17-triol, 13β-Methyl-1,3,5(10)-gonatriene-3,16α,17β-triol, 16α,17β-Estriol, 16α-Estriol, 16α-Hydroxyestradiol, 3,16α,17β-Estriol, 3,16α,17β-Trihydroxyestra-1,3,5(10)-triene, Aacifemine, Colpogyn, Destriol, Estra-1,3,5(10)-triene-3,16α,17β-triol, Estratriol, Follicular hormone hydrate, Gynasan, Holin V, Hormomed, Incurin, Klimax E, Klimoral, NSC 12169, OE3, Oekolp, Oestriol, Ortho-Gynest, Ovesterin, Ovestin, Ovo-Vinces, Theelol, Tridestrin, Trihydroxyestrin, Triovex
IUPAC Name (8R,9S,13S,14S,16R,17R)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,16,17-triol
SMILES C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1C[C@@H](O)[C@@H]2O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C18H24O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-17,19-21H,2,4,6-7,9H2,1H3/t13-,14-,15+,16-,17+,18+/m1/s1
Formula Weight 288.1725
Percent Purity >95%
Chemical Name or Material Estriol
Grade HPLC