Learn More
Thermo Scientific™ ES Series FMS Lab Freezers
Product Code. p-6065994 Shop All Thermo Scientific Products
Click to view available options
Capacity (Metric):
98 L
Plug Type:
EU
Unit Size:
Each
Product Code. 16681074
Brand: Thermo Scientific™ 98FAXVTS
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
The new Thermo Scientific™ ES Series FMS Lab Freezers are ideal for storing potentially flammable material in general laboratory settings. Household and commercial freezers are not suitable for such material: they have components in their electrical and refrigeration systems that can trigger explosions of flammable air-vapor mixtures inside the unit.
Reduce risk of accidental explosion and associated sample damage. The new Thermo Scientific™ ES Series FMS Lab Freezers are ideal for storing potentially flammable material in general laboratory settings. Household and commercial freezers are not suitable for such material: they have components in their electrical and refrigeration systems that can trigger explosions of flammable air-vapor mixtures inside the unit. These freezers have been constructed without components that could trigger an explosion or fire of the materials inside.
  • Integrated controller
  • Digital display
  • High- and low-temperature alarms
  • Door ajar alarm
  • Remote alarm contacts
  • Standard door locks
  • Access ports
  • CE/ATEX marked
  • Reversible doors
  • Hydrocarbn refrigerants
  • HCFC and CFC free insulation
Ordering Information: Available in 50Hz regions only; not available in North America and other 60Hz regions.
    Warranty varies by country, please contact us for details
TRUSTED_SUSTAINABILITY
Specifications
Type Corrosion-resistant, Spark-proof
Capacity (English) 3.5 cu. ft.
Capacity (Metric) 98 L
Temperature Range -10°C to -26°C
Refrigerant R600a
Door Style Hinged
Shelves 3
Cabinet Material Scratch-resistant Painted Steel
Insulation CFC-free, HCFC-free
Certifications/Compliance CE
Style Undercounter
Control Type Digital
Ports 1, 16 mm Side-Access Port
Defrost Automatic
Monitoring Options Alarm
Depth (Metric) Exterior 65.8 cm
Depth (Metric) Interior 44 cm
Height (Metric) Exterior 85 cm
Height (Metric) Interior 65 cm
Width (Metric) Exterior 59.9 cm
Width (Metric) Interior 42 cm
Plug Type EU
Voltage 230 V
Electrical Requirements 230 V, 50Hz
Frequency 50 Hz
Shipping Weight (Metric) 41.8 kg
Quantity Each
Show More Show Less
FMS units are not designed for use in Class I, Zone 0 or Zone 1 applications which require an explosion-proof unit.