Learn More
Erythrosin B, TRC
Specifications
Specifications
| Chemical Name or Material | Solvent Red 140 |
| CAS | 15905-32-5 |
| Molecular Formula | C20 H8 I4 O5 |
| Quantity | 10 g |
| Synonym | Fluorescein, 2',4',5',7'-tetraiodo- (8CI), 3',6'-Dihydroxy-2',4',5',7'-tetraiodospiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, 2',4',5',7'-Tetraiodofluorescein, C.I. 45430:2, C.I. Solvent Red 140, Erythrosin J, Erythrosine J, Erythrosine acid, Iodeosin, NSC 328781, Solvent Red 140, Tetraiodofluorescein |
| InChI Formula | InChI=1S/C20H8I4O5/c21-11-5-9-17(13(23)15(11)25)28-18-10(6-12(22)16(26)14(18)24)20(9)8-4-2-1-3-7(8)19(27)29-20/h1-6,25-26H |
| SMILES | Oc1c(I)cc2c(Oc3c(I)c(O)c(I)cc3C24OC(=O)c5ccccc45)c1I |
| IUPAC Name | 3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one |
| Molecular Weight (g/mol) | 835.89 |
| Formula Weight | 835.655 |