Learn More
BOLA Erlenmeyer Flasks
Click to view available options
Capacity (Metric):
50 mL
500 mL
Unit Size:
Each
Description
Thick-walled, with ground joint.
Specifications
Specifications
| Diameter (Metric) Base | 107 mm |
| Capacity (Metric) | 500 mL |
| Height (Metric) | 190 mm |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Graduated | No |
| Sterility | Non-sterile |
| Bottom Shape | Flat |