missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eriochrome™ Black T, Indicator, Reag. Ph. Eur. for metal titration, Honeywell Fluka™
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
CAS Max % | 100.0000% |
Linear Formula | C20H12N3NaO7S |
UN Number | NONH for all modes of transport |
Beilstein | 4121162 |
IUPAC Name | (4Z)-4-[(1-hydroxynaphthalen-2-yl)hydrazinylidene]-7-nitro-3-oxonaphthalene-1-sulfonic acid;sodium |
Formula Weight | 461.38 |
Grade | Ph. Eur. |
Packaging | Glass Bottle |
Suitability | Complies for indicator |
Quantity | 25 g |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'