missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ERCC8 (Human) Recombinant Protein
Human ERCC8 full-length ORF ( NP_000073.1, 1 a.a. - 396 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001161-P02.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MLGFLSARQTGLEDPLRLRRAESTRRVLGLELNKDRDVERIHGGGINTLDIEPVEGRYMLSGGSDGVIVLYDLENSSRQSYYTCKAVCSIGRDHPDVHRYSVETVQWYPHDTGMFTSSSFDKTLKVWDTNTLQTADVFNFEETVYSHHMSPVSTKHCLVAVGTRGPKVQLCDLKSGSCSHILQGHRQEILAVSWSPRYDYILATASADSRVKLWDVRRASGCLITLDQHNGKKSQAVESANTAHNGKVNGLCFTSDGLHLLTVGTDNRMRLWNSSNGENTLVNYGKVCNNSKKGLKFTVSCGCSSEFVFVPYGSTIAVYTVYSGEQITMLKGHYKTVDCCVFQSNFQELYSGSRDCNILAWVPSLYEPVPDDDETTTKSQLNPAFEDAWSSSDEEG
Specifications
NP_000073.1 | |
excision repair cross-complementing rodent repair deficiency, complementation group 8 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ERCC8 | |
GST |
1161 | |
Wheat germ expression system | |
10 μg | |
CKN1, CSA | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |