missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Erbstatin Analog
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Description
An inhibitor of the EGF receptor associated tyrosine kinase
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 1 mg |
| Description | Yellow |
| Content And Storage | -30°C to -10°C |
| For Use With (Application) | An inhibitor of the EGF receptor associated tyrosine kinase |
| Formula Weight | 194.19g/mol |
| Solubility | Soluble in methanol or 0.1M NaOH; Insoluble in water |
| Product Type | Signal Transduction Reagent |
| Form | Powder |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction