missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Equine IL-6 DuoSet ELISA

Product Code. 30001746
Click to view available options
Quantity:
1 Kit (for 15 Plates)
Unit Size:
Pack of 15
This item is not returnable. View return policy

Product Code. 30001746

Brand: R&D Systems™ DY1886

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For determination of natural and recombinant Equine IL-6 protein.

The Equine IL-6 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies equine IL-6.  

This DuoSet ELISA Development kit contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant equine Interleukin 6 (IL-6). The suggested diluent is suitable for the analysis of most cell culture supernate, serum, and plasma samples. Diluents for complex matrices, such as serum and plasma, should be evaluated prior to use in this DuoSet.
  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs
  • Economical alternative to complete kits
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 125.00 to 8000 pg/mL
Immunoassay Kit Format 96-well Plate
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Sufficient For Fifteen 96-well plates
Target Species Equine
For Use With (Application) ELISA
Gene ID (Entrez) 3569
Kit Contents Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)
Quantity 1 Kit (for 15 Plates)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Target IL-6
Show More Show Less

For Laboratory Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.