Learn More
MP Biomedicals™ Equine Donor Serum
Product Code. p-2625236 Shop All MP Biomedical Products
Click to view available options
Quantity:
100 mL
500 mL
Color:
Golden Brown
Unit Size:
100mL
500mL
Product Code. 11495035
Brand: MP Biomedicals™ 092921154
Specifications
Certifications/Compliance U.S. regulations
Color Golden Brown
Content And Storage -20°C
Species Horse
Form Liquid
Product Type Horse Serum
Quantity 500 mL
Sterility Sterile
Sterilization Method 0.2 μm filtration
For Use With (Application) Diagnostic assays
Hemoglobin Concentration ≤35mg/dL
Mycoplasma Detected No
Osmolality 273 to 301 mOsm/kg
pH 7.1 to 8
Protein Content 5.6 to 9.1 g/dL