Learn More
Equilin-d4, TRC
Product Code. 30379926
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30379926 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30379926 Supplier Toronto Research Chemicals Supplier No. TRCE5928021MG
 Restricted Item
Estimated Shipment: 12-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Equilin-2,4,16,16-d4
CAS 285979-79-5
Molecular Formula C18 2H4 H16 O2
Quantity 1 mg
Synonym Estra-1,3,5(10),7-tetraen-17-one-2,4,16,16-d4, 3-hydroxy-, 3-Hydroxyestra-1,3,5(10),7-tetraen-17-one-2,4,16,16-d4, Equilin-d4, Equilin-2,4,16,16-d4, Equilin-2,4,16,16-D4, 7-Dehydroestrone-d4, 7-Dehydroestrone-2,4,16,16-d4
InChI Formula InChI=1S/C18H20O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3-5,10,14,16,19H,2,6-9H2,1H3/t14-,16+,18+/m1/s1/i3D,7D2,10D
SMILES [2H]c1cc2[C@H]3CC[C@@]4(C)[C@@H](CC([2H])([2H])C4=O)C3=CCc2c([2H])c1O
IUPAC Name (9S,13S,14S)-2,4,16,16-tetradeuterio-3-hydroxy-13-methyl-6,9,11,12,14,15-hexahydrocyclopenta[a]phenanthren-17-one
Molecular Weight (g/mol) 272.37
Formula Weight 272.1714