Learn More
Equilin-2,4,16,16-d4, CDN
Product Code. 30320003
Change view
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30320003 0.05 g 0.05g
30311932 0.01 g 0.01g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30320003 Supplier CDN Supplier No. CDND41860.05G
 Restricted Item
Estimated Shipment: 10-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Equilin-2,4,16,16-d4
CAS 285979-79-5
Molecular Formula C18 2H4 H16 O2
Quantity 0.05 g
Synonym Estra-1,3,5(10),7-tetraen-17-one-2,4,16,16-d4, 3-hydroxy-, 3-Hydroxyestra-1,3,5(10),7-tetraen-17-one-2,4,16,16-d4, Equilin-d4, Equilin-2,4,16,16-d4, Equilin-2,4,16,16-D4, 7-Dehydroestrone-d4, 7-Dehydroestrone-2,4,16,16-d4
InChI Formula InChI=1 S/C18H20O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3-5,10,14,16,19 H,2,6-9H2,1H3/t14-,16+,18+/m1/s1/i3D,7D2,10 D
SMILES [2 H]c1cc2[C@H]3 CC[C@@]4(C)[C@@H](CC([2 H])([2 H])C4=O)C3=CCc2c([2 H])c1O
IUPAC Name (9 S,13 S,14 S)-2,4,16,16-tetradeuterio-3-hydroxy-13-methyl-6,9,11,12,14,15-hexahydrocyclopenta[a]phenanthren-17-one
Molecular Weight (g/mol) 272.37
Formula Weight 272.1714 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity