Learn More
- Home
- Products
- Facility Safety and Maintenance
- Labels and Label Makers
- Custom Labels
- Brady™ EPREP Labels for M710 Printer
Brady™ EPREP Labels for M710 Printer
Product Code. 17897924 Shop All Brady Products
Change view
Click to view available options
Color:
Black
Green
Red
Silver
White
Yellow
Dimensions (L x W):
7.95 x 27 mm
12.45 x 27 mm
15 x 27 mm
15 x 45 mm
18 x 27 mm
18 x 35 mm
19 x 48 mm
40 x 30 mm
Width (English) Printable Area:
27 mm
30 mm
35 mm
45 mm
48 mm
Unit Size:
Pack of 100
Pack of 125
Pack of 150
Pack of 250
Pack of 500
24 product options available for selection
| Product Code. | Color | Dimensions (L x W) | Width (English) Printable Area | Unit Size |
|---|---|---|---|---|
| 17897924 | Silver | 18 x 35 mm | 35 mm | Pack of 100 |
| 17848024 | White | 7.95 x 27 mm | 27 mm | Pack of 250 |
| 17848084 | White | 12.45 x 27 mm | 27 mm | Pack of 250 |
| 17868084 | White | 15 x 27 mm | 27 mm | Pack of 150 |
| 17827954 | White | 15 x 45 mm | 45 mm | Pack of 500 |
| 17888004 | White | 18 x 27 mm | 27 mm | Pack of 150 |
| 17898024 | White | 18 x 35 mm | 35 mm | Pack of 100 |
| 17847934 | White | 19 x 48 mm | 48 mm | Pack of 125 |
| 17898004 | White | 40 x 30 mm | 30 mm | Pack of 100 |
| 17858084 | Black | 15 x 45 mm | 45 mm | Pack of 100 |
| 17887924 | Black | 40 x 30 mm | 30 mm | Pack of 100 |
| 17828044 | Silver | 7.95 x 27 mm | 27 mm | Pack of 250 |
| 17867924 | Silver | 12.45 x 27 mm | 27 mm | Pack of 250 |
| 17838084 | Silver | 15 x 27 mm | 27 mm | Pack of 150 |
| 17818084 | Silver | 15 x 45 mm | 45 mm | Pack of 500 |
| 17868004 | Silver | 18 x 27 mm | 27 mm | Pack of 150 |
| 17858014 | Silver | 19 x 48 mm | 48 mm | Pack of 125 |
| 17818044 | Silver | 40 x 30 mm | 30 mm | Pack of 100 |
| 17818014 | Red | 15 x 45 mm | 45 mm | Pack of 100 |
| 17878004 | Red | 40 x 30 mm | 30 mm | Pack of 100 |
| 17807934 | Yellow | 15 x 45 mm | 45 mm | Pack of 100 |
| 17868014 | Yellow | 18 x 35 mm | 35 mm | Pack of 100 |
| 17807864 | Yellow | 40 x 30 mm | 30 mm | Pack of 100 |
| 17877924 | Green | 40 x 30 mm | 30 mm | Pack of 100 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Aggressive adhesive and solvent resistance makes them an ideal replacement for engraved tags
- Service temperature of -40°C to +100°C
Specifications
Specifications
| Adhesive Type | Permanent Acrylic, Foam-backed |
| Coating | None |
| Color | Silver |
| Finish | Gloss |
| Format | Label |
| Material | Polyester |
| Printable Sides | Single-side Printable |
| Print Technology | Thermal Transfer |
| Product Type | Label |
| Sterility | Non-sterile |
| For Use With (Application) | Data and Telecommunications, Electrical Labeling, General Identification |
| For Use With (Equipment) | M710, BMP71 |
| Labels per Row | 1 Label |
| Min. Operating Temperature (Metric) | -40°C |
| Quantity | 100 Labels |
| Shape | Rectangle |
| Temperature Range (English) | -40°F to +212°F |
| Temperature Range (Metric) | -40°C to +100°C |
| Thickness (English) | 0.0256 in. |
| Thickness (Metric) | 0.65 mm |
| Dimensions (L x W) | 18 x 35 mm |
| Length (English) | 0.71 in. |
| Width (English) | 1.38 in. |
| Width (Metric) | 35 mm |
| Width (English) Printable Area | 35 mm |
| Alternate Part Number | M7-7-7593-SL |
| Show More Show Less |
Product Title
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?