Learn More
Eppendorf™ Sticky Pad Adapter Kit for Biological Shakers
Product Code. 11339874
Click to view available options
For Use With (Equipment):
Biological shaker
Unit Size:
Each
Product Code. 11339874
Brand: Eppendorf™ M12509504
Only null left
Add to basket
Add to basket
Alternative to flask clamps for securing vessels to a shaker platform
  • Ideal for containers such as bottles and dishes that cannot be held with conventional flask clamps
  • 45.72 × 15.24cm (18 × 6 in.)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Sticky Pad Adapter Kit
Dimensions (L x W) 18 x 6 in. (45.72 x 15.24 cm)
Length (English) 18 in.
Length (Metric) 45.72 cm
Width (English) 6 in.
Width (Metric) 15.24 cm
For Use With (Equipment) Biological shaker
Includes Two anodized Aluminum sheets