Learn More
Eppendorf™ Combitips™ Advanced Tips for Multipette™ Multi-dispensers - Eppendorf Quality
Product Code. 12664587 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
0.1 mL
0.2 mL
0.5 mL
1.0 mL
2.5 mL
5.0 mL
10 mL
25 mL
50 mL
Color Code:
Blue
Green
Light Blue
Light Gray
Orange
Red
Violet
White
Yellow
Unit Size:
Pack of 100
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color Code Unit Size
12664587 2.5 mL Green Pack of 100
12644587 0.1 mL White Pack of 100
12684587 0.2 mL Light Blue Pack of 100
12674587 0.5 mL Violet Pack of 100
12654587 1.0 mL Yellow Pack of 100
12694587 5.0 mL Blue Pack of 100
12657636 10 mL Orange Pack of 100
12604597 25 mL Red Pack of 100
12624597 50 mL Light Gray Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 12664587 Supplier Eppendorf™ Supplier No. 0030089448
  • ONLINE EXCLUSIVE PRICE
    121.00€
    112.00€ / Pack of 100
    Save 9.00€
    7% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Bulk Quote Request
Eppendorf's consumable of choice for all Eppendorf Multipette™ multi-dispensers. Available in 9 volume sizes (0.1 to 50 mL), Combitips advanced in 'Eppendorf Quality' dispense in increments as low as 100 mL and are ideally suited for a variety of standard lab applications. Also available in purity grades PCR clean, Biopur™ or Forensic DNA Grade.
Use these tips for comfortable handling while preventing damage to gloves. Eppendorf Combitips advanced™ Standard Pipetter Tips are optimized to meet the increased requirements of a modern laboratory, and offered in three different purity grades to support any application.
  • Combitips advanced are the ideal system components for all Eppendorf Multipettes
  • The 9 volume sizes (0.1 mL – 50 mL) offer a maximum range of dispensing volumes with increments as low as 100 nanoliters depending on the Combitip used
  • Elongated Combitips in volumes 2.5 mL, 5.0 mL and 10 mL make it possible to reach the bottom of the most common laboratory vessels
  • An individual color coding facilitates the quick identification of the desired Combitips
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 2.5 mL
Filtered Non-filtered
Sterility Non-sterile
Color Colorless
Autoclavable Not Autoclavable
Clean Claims Eppendorf Quality (Non-sterile)
Format Bulk, Bagged
Tip Style Extended Length, Positive Displacement
Packaging 100 Pk. (4 Bags x 25 Pk.)
Product Type Pipette Tips
Color Code Green
Disposable Single-use
For Use With (Equipment) Positive Displacement System
Graduated Yes
Product Line CombiTips Advanced
Material Polypropylene
Fit for Purpose Certified Quality