missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EpiCult™-C Human Medium Kit
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
- EpiCult™-C Medium (Human) is a serum-free culture medium optimized for the short-term culture of human mammary luminal and myoepithelial cells
- This medium is also ideal for the enzymatic dissociation of human mammary tissue when supplemented with Collagenase/Hyaluronidase (Catalog #07912)
- This kit contains EpiCult™-C Basal Medium (Human) and EpiCult™-C Proliferation Supplement (Human)
- Addition of Hydrocortisone Stock Solution (Catalog #07925) is required
Specifications
Specifications
| Cell Type | Mammary Cells |
| For Use With (Application) | Maintenance |
| Quantity | 500 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction