missing translation for 'onlineSavingsMsg'
Learn More

epi-Sesamin Monocatechol, TRC

Product Code. 30427539
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427539 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30427539 Supplier Toronto Research Chemicals Supplier No. TRCS28051510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material epi-Sesamin Monocatechol
CAS 1105568-81-7
Molecular Formula C19 H18 O6
Quantity 10 mg
Synonym 4-[(3aR,6aR)-4-(1,3-Benzodioxol-5-yl)tetrahydro-1H,3H-furo[3,4-c]furan-1-yl]-1,2-benzenediol
InChI Formula InChI=1S/C19H18O6/c20-14-3-1-10(5-15(14)21)18-12-7-23-19(13(12)8-22-18)11-2-4-16-17(6-11)25-9-24-16/h1-6,12-13,18-21H,7-9H2/t12-,13-,18+,19+/m0/s1
SMILES Oc1ccc(cc1O)[C@H]2OC[C@H]3[C@@H]2CO[C@@H]3c4ccc5OCOc5c4
IUPAC Name 4-[(3S,3aR,6S,6aR)-6-(1,3-benzodioxol-5-yl)-1,3,3a,4,6,6a-hexahydrofuro[3,4-c]furan-3-yl]benzene-1,2-diol
Molecular Weight (g/mol) 342.34
Formula Weight 342.1103
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.