Learn More
Memmert™ Entry Port Right Centre/Centre
Product Code. 17160105 Shop All Memmert Products
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP260eco
IPP410eco
IPP750eco
missing translation for 'unitSize'
Each
Product Code. 17160105
missing translation for 'mfr': Memmert™ IPP110ecoF2
Only null left
Add to basket
Add to basket
For introducing connections at the side and can be closed by flap
  • Standard position right centre/centre
  • Not for model sizes 110 and 260 with light module
  • Not for model size 110 with light module
Specifications
Color Clear
For Use With (Equipment) IPP110eco
Diameter (Metric) 23 mm