Learn More
Memmert Entry port 23mm, flap & stopper closure, standardposition left side centre/centre, for ICH260
Brand:  Memmert ICH260F0
Entry port 23mm, flap & stopper closure, standardposition left side centre/centre, for ICH260
Product Code. 16336657
  • ONLINE EXCLUSIVE PRICE
    226.00€
    218.00€ / Each
    Save 8.00€
    3% Off
Only null left
Estimated Shipment: 17-03-2026