Learn More
Memmert Entry port 23mm, flap & stopper closure, standardposition left side centre/centre, for ICH110
Brand:  Memmert ICH110F0
Entry port 23mm, flap & stopper closure, standardposition left side centre/centre, for ICH110
Product Code. 16315237
  • ONLINE EXCLUSIVE PRICE
    240.00€
    218.00€ / Each
    Save 22.00€
    9% Off
Only null left
Estimated Shipment: 16-03-2026