Learn More
Memmert™ Entry Port 23 mm, Standard Position Left Side Centre/Top, For Ich750
Shop All Memmert Products
Click to view available options
Diameter (Metric):
23 mm
Unit Size:
Each
Description
- Entry port with clear diameter
- For Memmert Ovens
Specifications
Specifications
| For Use With (Equipment) | Models 30, 55, 75, 110, 160, 260, 450, 750, 1060 |
| Material | Stainless Steel |
| Type | Entry Port |
| Diameter (Metric) | 23 mm |