missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Endothelin 3 (human, rat)

Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
Endothelin 3 (human, rat) is an endogenous neuropeptide and potent vasoconstrictor. Displays selectivity for the putative ETC endothelin receptor.
Specifications
Specifications
| Description | Endothelin 3 (human, rat) |
| Molecular Weight (g/mol) | 2643 |
| Product Type | Endothelin 3 (human, rat) |
| Quantity | 100 μg |
| Sequence | Cys-Thr-Cys-Phe-Thr-Tyr-Lys-Asp-Lys-Glu-Cys-Val-Tyr-Tyr-Cys-His-Leu-Asp-lle-Ile-Trp |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction