Learn More
R&D Systems™ Endothelin 1 (human, porcine)
Product Code. 18718896
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18718896 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18718896 Supplier R&D Systems™ Supplier No. 1160/100U
  • ONLINE EXCLUSIVE PRICE
    312.53€
    281.40€ / 100µg
    Save 31.13€
    9% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
A Tocris product, part of R&D Systems. Potent vasoconstrictor peptide
Endothelin 1 (human, porcine) is an endogenous potent vasoconstrictor peptide that is an agonist at ETA and ETB receptors. Modulates vascular tone.
TRUSTED_SUSTAINABILITY
Specifications
Product Line Tocris
Quantity 100 μg