missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ENDOG (Human) Recombinant Protein
Human ENDOG full-length ORF ( AAH04922.1, 50 a.a. - 297 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002021-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: ELPPVPGGPRGPGELAKYGLPGLAQLKSRESYVLCYDPRTRGALWVVEQLRPERLRGDGDRRECDFREDDSVHAYHRATNADYRGSGFDRGHLAAAANHRWSQKAMDDTFYLSNVAPQVPHLNQNAWNNLEKYSRSLTRSYQNVYVCTGPLFLPRTEADGKSYVKYQVIGKNHVAVPTHFFKVLILEAAGGQIELRTYVMPNAPVDEAIPLERFLVPIESIERASGLLFVPNILARAGSLKAITAGSK
Specifications
AAH04922.1 | |
endonuclease G | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ENDOG | |
GST |
2021 | |
Wheat germ expression system | |
25 μg | |
FLJ27463 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |