Learn More
Ansell ENCORE™ Latex Ortho Surgical Gloves
Product Code. p-7250183 Shop All Ansell Products
Click to view available options
Size:
6
6.5
7
7.5
8
8.5
9
Unit Size:
50 pairs
This product contains natural rubber latex, which may cause allergic reactions.
Product Code. 17833622
Brand: Ansell 330106070
Only null left
Add to basket
Add to basket
This product contains natural rubber latex, which may cause allergic reactions.
Latex surgical gloves with extra-thick glove formulation to withstand the rigors of surgery, especially where power tools and bone fragments are present.
  • ASTM D3577, ASTM D6978 certified
  • 50% thicker on average than Ansell's standard powder-free latex surgical gloves, providing outstanding protection
  • Heavy-textured finish on fingers and palm for superior grip for dry and damp applications
  • DERMASHIELD™ Technology internal coating facilitates dry and damp donning as well as intra-operative changes
  • For use in rigorous surgical procedures
Specifications
Certifications/Compliance ASTM D3577, ASTM D6978, ISO 10282
Certifications/Compliance (Europe) EN 16523-1, EN 374:2003, EN 388:2003, EN 388:2016 +A1:2018, EN 420:2003 + A1:2009, EN 455 1-4, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, EN ISO 374-1:2016, EN 374-2 and -4, EN ISO 374-5:2016, EN 388:2016
Accelerator Free No
Applications Healthcare
AQL 0.65
Color Brown
Disposable Single-use
Exterior Finish Fully Textured
Finger Thickness (English) 12.8 ±1.18 mil
Finger Thickness (Metric) 0.33 ±0.03 mm
Hand Specific
Laboratory/Cleanroom Use No
Latex Free No
Material Latex
Palm Thickness (English) 12.01 ±1.18 mil
Palm Thickness (Metric) 0.31 ±0.03 mm
Powdered Powder Free
Size 7
Sterility Sterile
Thickness (English) 12.01 ±1.18 mil
Thickness (Metric) 0.31 ±0.03 mm
Model ENCORE-330106
Length (English) 11.9 in.
Length (Metric) 301 mm