missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ EMPA
GREENER_CHOICE

Product Code. 30001565
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001565 50 mg 50mg
30001550 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001565 Supplier Tocris Bioscience™ Supplier No. 4558/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Highly potent and selective OX2 antagonist; brain penetrant

Highly potent, selective OX2 receptor antagonist (IC50 values are 2.3 nM and 1900 nM for OX2 and OX1 respectively). Displays negligible or no inhibition of a panel of 80 receptors. Blocks orexin-B- and orexin-A-invoked calcium mobilization in hOX2-expressing CHO cells (IC50 values are 7.9 nM and 8.8 nM respectively); reverses orexin-B-induced hyperlocomotion in mice. Brain penetrant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 680590-49-2
Molecular Formula C23H26N4O4S

Specifications

Quantity 50 mg
Formula Weight 454.54
Purity >98%
Molecular Weight (g/mol) 454.545
Chemical Name or Material EMPA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.