Learn More
Merck Accessories and Replacement Parts In-Line Stainless Steel Filter Holders 47 mm
Accessories for filter holders for sample preparation.
Brand:  Merck XX4404702
517.79 EUR valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10029810
  • 560.00€ / Each
Estimated Shipment: 22-08-2024
Description
XX4204709, XX4304701, XX4404703, 5614, and 00907P are replacement parts for Filter Holder XX4404700
SS Filter, Depends on filter type used.
Specifications
Specifications
Not Autoclavable
47 mm high Pressure SS filter holder
Non-sterile
PTFE-coated HP Support Screen
PTFE Coated Stainless Steel
47 mm