Learn More
Merck Luminata™ Starter Package
Product Code. 12541289 Shop All Merck Products
Change view
Click to view available options
Type:
Luminata Starter Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
12541289 Luminata Starter Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12541289 Supplier Merck Supplier No. WBLUM0100
  • ONLINE EXCLUSIVE PRICE
    412.00€
    383.00€ / Pack of 1
    Save 29.00€
    7% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Western blot packages with unbelievable savings! Save time and money by choosing the package that best serves your needs!
Samples of all three Luminata Western HRP substrates for antibodies of all affinities
  • Luminata Classico Western HRP Substrate, 100mL (WBLUC0100)
  • Luminata Crescendo Western HRP Substrate, 100mL (WBLUR0100)
  • Luminata Forte Western HRP Substrate, 100mL (WBLUF0100)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Western blotting, binding assays, amino acid analysis, N-terminal protein sequencing, dot/slot blotting, glycoprotein visualization, lipopolysaccharide analysis.
For Use With (Equipment) Snap i.d.™ Protein Detection System
Type Luminata Starter Kit
Includes Luminata Classico Western HRP Substrate, 100mL (WBLUC0100)
Luminata Crescendo Western HRP Substrate, 100mL (WBLUR0100)
Luminata Forte Western HRP Substrate, 100mL (WBLUF0100)