Learn More
Merck PureProteome™ Nickel Magnetic Beads
Click to view available options
Quantity:
10 mL
2 x 1 mL
Unit Size:
Each
Pack of 2
Description
- No lysate clearing—crude cell lysates are processed by magnetic beads
- Fast processing time—entire protocol is completed in less than one hour
- High capacity—bind 5 to 15mg of protein per mL of settled beads
- High affinity—binding can take place in buffer containing <5mM EDTA
- Flexible scale—varying the bead quantity allows different protein yields
- Flexible elution—native or denatured proteins
- Automatable—hands-free screening process
Specifications
Specifications
| Binding Property | 1 to 5.5mg/mL |
| Content And Storage | 2°C to 8°C |
| Particle Size | 10 μm |
| Quantity | 2 x 1 mL |
| For Use With (Application) | Purification of His-tagged recombinant Protein |
| Type | Magnetic Affinity Bead |
Safety and Handling
Product Identifier
- HisPur Ni-NTA Magnetic Beads
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P264-Wash skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Sulfuric acid, nickel(2+) salt