Learn More
Merck PureProteome™ Carboxy FlexiBind Magnetic Bead System
Shop All Merck Products
Click to view available options
Quantity:
10 mL
Particle Size:
1μm
2.5μm
Unit Size:
Each
Description
Custom affinity bead generation for binding to target ligand.
- Advanced chemistry combined with high surface area provides more binding sites for proteins
- Particles visible as they adhere to side of tube for quick and easy aspiration and complete buffer removal
- Beads are immobilized in seconds
- Increased kinetics of bead-protein binding enables shorter incubations
- Significantly more affordable then competitive magnetic beads
Research Use Only
Specifications
Specifications
| Product Type | Carboxy flexiBind Magnetic bead system |
| Quantity | 10 mL |
| Particle Size | 1μm |
| Content And Storage | 2° to 8°C |
| For Use With (Application) | Immunoprecipitations, purifying Nucleic Acids, isolating cells/organelles, Protein-protein interactions |
| Matrix | Magnetic Beads |