Learn More
Merck Accessories for Patch Test Kit
Product Code. 10547651 Shop All Merck Products
Change view
Click to view available options
Type:
Filter Holder
Filter Holder Support with Stainless Steel Base
Funnel Locking Ring
Locking Ring Gasket
Naval Fluid Color Guide
Sample Bottle
Solvent Dispensing Bottle
Tubing for vacuum
Unit Size:
Each
Pack of 5
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10547651 Funnel Locking Ring Each
10707291 Locking Ring Gasket Pack of 5
10089840 Filter Holder Each
10717481 Solvent Dispensing Bottle Each
10747671 Sample Bottle Each
10032473 Filter Holder Support with Stainless Steel Base Each
10667493 Naval Fluid Color Guide Each
10192061 Tubing for vacuum Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10547651 Supplier Merck Supplier No. XX2004701
  • ONLINE EXCLUSIVE PRICE
    189.00€
    185.00€ / Pack of 1
    Save 4.00€
    2% Off
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
Accessories for filter holders for sample preparation.
The MilliporeSigma Patch Test Kit is a complete solution for field-based collection and analysis of hydrocarbon-based hydraulic fluids, bulk chemicals, boiler water, and lubricating oils. It allows the detection of significant changes in cleanliness through dependable, sensitive, colorimetric-based analysis.- Monitors particulate contamination rapidly in hydrocarbon-based hydraulic fluids, bulk chemicals, boiler water and lubricating oils - Filter color rating and particle assessment scales correspond to recognized standard contamination levels - Detects significant changes in cleanliness through dependable and sensitive test results - Comes with filters for 100 tests Features & Benefits:•Contains enough expendable materials for 100 tests •Contained within a lightweight, easy-to-carry case •Filter color rating and particle assessment scales correspond to recognized standard contamination levels Applications:In-Field Contamination Analysis of Hydrocarbon-Based Hydraulic Fluids, Water, Lubricating Oils and Related ProductsKit Catalog Number XX6504730 Patch Test KitComponentsPD1504700 PetriSlides for contamination analysisSMWP02500 MF-Millipore Membrane Filter, 5.0 μm, 25 mm, white, plain, 100SMWP04700 MF-Millipore Membrane Filter, 5.0 μm, 47 mm, white, plain, 100XX6200006P Filter forceps, blunt end, stainless steelXX6200035 Hand Vacuum Pump, stainless steelXX6300120 Patch Test Filter Holder AssemblyXX6504704 Solvent Dispensing BottleXX6504707 Swinnex 25 mm with back-pressure screenXX6504709 PVC Bottle, 120 mLXX6504710 Tubing for vacuum, 1/8 in., fluoroelastomerXX6504713 Naval Fluid Color Guide 1Replacement Parts Clear Sorting & FilteringXX2004701 Funnel Locking RingXX2004703 Gasket, PTFEXX4004714 Locking Ring Gasket, PTFEXX6300123 O-ring, 2-233, Buna-NXX6504708 Stainless Steel Holder Support with base
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Patch Test Kit
Material Anodized Aluminum
Type Funnel Locking Ring