Learn More
Merck Millicell™ EZ Slides
Product Code. 11795254 Shop All Merck Products
Change view
Click to view available options
Wells:
4
8
Quantity:
16 Pk.
96 Pk.
Unit Size:
Pack of 16
Pack of 96
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wells Quantity Unit Size
11795254 4 96 Pk. Pack of 96
11765254 8 16 Pk. Pack of 16
11755254 4 16 Pk. Pack of 16
11705264 8 96 Pk. Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11795254 Supplier Merck Supplier No. PEZGS0496
  • ONLINE EXCLUSIVE PRICE
    967.00€
    917.00€ / Pack of 96
    Save 50.00€
    5% Off
Estimated Shipment: 19-11-2026
Culture, fix, stain, and view cells all in one device
  • Polypropylene wells are resistant to most fixatives—cells may be fixed and stained with wells still attached
  • Breakable tabs for easy well removal—no tools required to remove wells from slide
  • No adhesive used to attach wells to slide
  • No leaky wells
  • Raised corner ridges on lid facilitate stacking of multiple units in the incubator
  • Noncytotoxic and nonpyrogenic
  • Sterile
  • Slide holder (available separately) protects slide from damage
TRUSTED_SUSTAINABILITY
Specifications
Type EZ Slide
Wells 4
Color Clear
Well Material Polypropylene
Culture Area 1.7 cm2/well
Lid Material Polystyrene
Material Glass
Volume (Metric) Working 0.5 to 1.7 mL/Well
For Use With (Application) Simplify your cell analysis by using the Millicell EZ SLIDE glass slide to culture, fix, stain and view your sample all in one device
Sterility Sterile
Quantity 96 Pk.