Learn More
Merck Millicell-ERS2 Volt-Ohm Meter and accessories
Product Code. 11760363 Shop All Merck Products
Click to view available options
Product Type:
Adjustable Electrode
Fixed Test Electrode
Millicell Ers 2V Ohm Meter
Specialized Electrode
Unit Size:
Each
Product Code. 11760363
Brand: Merck MERSSTX04
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Includes
Millicell-ERS2 comes with meter, battery, STX01 electrode, 1,000 Ω test electrode, battery charger. Specialized electrodes for Millicell-96 well plate are available (Cat. No. MERSSTX00).
Measures Trans Epithelial Electrical Resistance (TEER) of epithelial cells in culture
Provides qualitative measurement of cell monolayer health and quantative measures of confluency. Designed exclusively for use with cells cultured in Millicell™ Culture Plate Inserts.
Utilizes AC current for membrane resistance measurements, producing a zero net charge on cells and eliminating adverse DC current effects on cell membranes. A silver/silver chloride (Ag/AgCI) pellet on each electrode tip measures voltage. Due to the small size of the electrodes, the user can easily measure transepithelial voltage and the resistance of cells grown on microporous membranes.
Warranty
Electrodes and batteries are warranted for 30 days only from the date on which the customer receives these items
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fixed Test Electrode
For Use With (Equipment) Millicell-96 Cell Culture Insert Plates
Length (English) 7.3 in.
Length (Metric) 19 cm
For Use With (Application) To provide qualitative measurement of cell monolayer health and quantative measures of confluency
Height (English) 2.3 in.
Height (Metric) 6 cm
Hertz 12.5 Hz
Humidity 0 to 90%
Includes Millicell-ers2 comes with meter, battery, stx01 electrode, 1000w test electrode, battery charger, specialized electrodes for millicell-96 well plate
Run Time 8 to 10 hr.
Temperature Range (English) 50° to 100°F
Temperature Range (Metric) 10° to 38°C
Weight (English) 3 lb.
Weight (Metric) 1.4 kg
Width (Metric) 11 cm