Learn More
Merck Integrity Test Kit for Small Volume Devices
For small volume devices
Brand:  Merck SLTEST000
371.00 EUR valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.10000kg
Product Code. 10762161
  • 399.00€ / Each
Estimated Shipment: 24-05-2024
Description
  • Convenient bubble-point testing
  • For vented and non-vented devices
  • Tests wide range of pore sizes and membrane types
  • Millipore membrane filter units can be integrity-tested using the bubble point test method. Small, syringe-driven membrane filter units with 15 cm2 filtration area, such as Millex and Sterivex units, can be conveniently tested using the syringe-style integrity tester.
    Specifications
    Specifications
    5 μm
    15 cm2
    Integrity Tester