missing translation for 'onlineSavingsMsg'
Learn More

Merck Chemicals Enzyme Free Cell Dissociation Solution Hank's Based (1x)

Product Code. p-5235526
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 17646205

Brand: Merck Chemicals S004C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hank's based formulation of chelating agents and agents to stabilize their activity on cells

Enzyme-Free Cell Dissociation Solution is a Hank's based formulation of chelating agents and agents to stabilize their activity on cells. This reagent does NOT contain porcine trypsin or any other protein. It works significantly faster than 0.5mM EDTA. In fact, it works as quickly as trypsin on most cells. Most importantly, it preserves the structural and functional integrity of cell surface proteins and does not have the cytotoxic effects sometimes associated with 0.5mm EDTA. It can be used to dissociate primary cells, tissues, and tumors with an increased plating efficiency.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Formulation Chelating and stabilizing agents
Quantity 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.