Learn More
Merck Empty Monitor Cassette, 25 mm, Type A
Product Code. 10608091 Shop All Merck Products
Change view
Click to view available options
For Use With (Equipment):
EMD Millipore Aerosol Analysis Monitor (25 mm)
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10608091 EMD Millipore Aerosol Analysis Monitor (25 mm) Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10608091 Supplier Merck Supplier No. M000025A0
  • ONLINE EXCLUSIVE PRICE
    453.00€
    415.99€ / Pack of 50
    Save 37.01€
    8% Off
Estimated Shipment: 10-12-2026
Sample collection and particle analysis
Accessory for 3-piece PS monitor, Type A
Order Info
Bulk pack.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Environmental Test Kits or Supplies
Material Polystyrene
Connector Types Female Luer slip, requires tubing adapter XX62 000 05
For Use With (Equipment) EMD Millipore Aerosol Analysis Monitor (25 mm)
For Use With (Application) Sample collection, Particle analysis, Environmental analysis
Includes Empty Monitor Cassette, 25  mm, Type A