Learn More
Merck Millipore Chemicon™ Accutase™ Cell Detachment Solution
useful for creating single cell suspensions from clumped cell cultures for accurate cell counting, detachment of cells from primary tissue.
Brand: Merck Millipore SCR005
This item is not returnable.
View return policy
Description
Useful for the routine detachment of cells from standard tissue culture plasticware and adhesion coated plasticware, including SmartPlastic™. ACCUTASE™ does not contain mammalian or bacterial derived products.